missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Src (Y419) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems AF2685-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Src Polyclonal specifically detects Src in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Especificaciones
| Src (p Tyr419) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SRC | |
| Phosphopeptide containing human Src Y419 site | |
| 25 μg | |
| Primary | |
| Detects human Src family members when phosphorylated at sites corresponding to Y419 of human Src in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ASV, c-Src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, RSVgp4, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6714 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
For Research Use Only