missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-EGFR/ErbB1 (Y1068) Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18715249
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18715249 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18715249 Proveedor R&D Systems N.º de proveedor IC3570U100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno EGFR
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 338324
Concentración 0.2 mg/mL
Conjugado Alexa Fluor 350
Dilución Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Alias de gen avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Símbolos de los genes EGFR
Especie del huésped Mouse
Inmunógeno Phosphopeptide containing human EGF R Y1068 site
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1956
Especificidad de la prueba Detects human EGF R phosphorylated at Y1068.
Especies diana Human
Contenido y almacenamiento Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.