missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Axl (Y779) Alexa Fluor 750-conjugated Antibody, R&D Systems™

Description
Axl Monoclonal specifically detects Axl in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
| Antígeno | Axl |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 713610 |
| Concentración | 0.2 mg/mL |
| Conjugado | Alexa Fluor 750 |
| Dilución | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulación | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Alias de gen | AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL |
| Símbolos de los genes | AXL |
| Show More |
For Research Use Only (RUO)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?