Learn More
Human PD-1 Antibody, R&D Systems™
Código de producto. 30306010
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
30306010 25 μg 25 microgramos
30306035 100 μg 100 microgramos
2 options
Código de producto. 30306010
Marca: R&D Systems MAB11638SP
Rabbit Monoclonal Antibody
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PD-1
Aplicaciones Immunohistochemistry, Multiplex Immunofluorescence
Clasificación Monoclonal
Clon 2705E
Conjugado Unconjugated
Dilución Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
N.º de referencia del gen Q15116
Alias de gen CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Especie del huésped Rabbit
Inmunógeno Synthetic Peptide, Accession # Q15116
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5133
Reconstitución Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.