missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Descripción
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | PD-1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1026519 |
| Conjugado | Alexa Fluor 700 |
| Dilución | Flow Cytometry 0.25-1 ug/106 cells |
| Formulación | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Alias de gen | CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Especie del huésped | Mouse |
| Inmunógeno | Synthetic peptide containing human PD-1 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?