missing translation for 'onlineSavingsMsg'
Learn More

Human Pax4 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18708417
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18708417 25 μg 25 microgramos
18038711 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18708417 Proveedor R&D Systems N.º de proveedor AF2614SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Goat Polyclonal Antibody has been used in 2 publications

Pax4 Polyclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Pax4
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen NP_006184
Alias de gen KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4
Símbolos de los genes PAX4
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human Pax4 Met1-Glu343 Accession # NP_006184
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5078
Especificidad de la prueba Detects human Pax4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax3 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.