missing translation for 'onlineSavingsMsg'
Learn More

Human Pax4 Antibody, R&D Systems™

Código de producto. 18038711
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
Código de producto. Cantidad unitSize
18038711 100 μg 100 microgramos
18708417 25 μg 25 microgramos
2 options
This item is not returnable. View return policy

Product Code. 18038711

Brand: R&D Systems AF2614

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Pax4 Polyclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Pax4
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
N.º de referencia del gen NP_006184
Alias de gen KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4
Símbolos de los genes PAX4
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human Pax4 Met1-Glu343 Accession # NP_006184
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5078
Especificidad de la prueba Detects human Pax4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax3 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.