Learn More
Human P2X7/P2RX7 Antibody, R&D Systems™
Descripción
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | P2X7/P2RX7 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1058613 |
| Conjugado | Unconjugated |
| Dilución | Flow Cytometry 0.25 ug/10^6 cells |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| N.º de referencia del gen | Q99572 |
| Alias de gen | ATP receptor, MGC20089, P2RX7, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A |
| Especie del huésped | Mouse |
| Mostrar más |