Learn More
Invitrogen™ Human Oxidized LDL ELISA Kit
Click to view available options
Cantidad:
96 pruebas
Tamaño de la unidad:
paquete de 96
Descripción
Low density lipoprotein (LDL) oxidative modification is one of the major processes involved in atherosclerosis along with other degenerative disorders. Oxidized LDL is preferentially accumulated by macrophages which become so calledfoam cells within the atheromatous plaque and contributes to the accumulation of cholesterol, typical of atherosclerosis.
Especificaciones
Especificaciones
| Intervalo del ensayo | 62.5 to 4000 pg/mL |
| Sensibilidad del ensayo | 37.5 pg/mL |
| Conjugado | HRP |
| Tipo de producto | ELISA |
| Tipo de muestra | Plasma, Serum |
| Para utilizar con (equipo) | Lector de microplacas colorimétrico |
| CV interensayo | <10% |
| CV intraensayo | <10 |
| Contenido del kit | Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer |
| Etiqueta o tinte | HRP |
| Mostrar más |