missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40/TNFRSF4 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marca: R&D Systems BAF3388
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
OX40/TNFRSF4 Polyclonal specifically detects OX40/TNFRSF4 in Human samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones
| OX40/TNFRSF4 | |
| Polyclonal | |
| Biotin | |
| P43489 | |
| TNFRSF4 | |
| Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4 Leu29-Ala216 Accession # P43489 | |
| 50 μg | |
| Primary | |
| Detects human OX40/TNFRSF4 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse OX40 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7293 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido