missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Otolin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB8045-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Otolin-1 Monoclonal specifically detects Otolin-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Especificaciones
| Otolin-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| A6NHN0 | |
| OTOL1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Otolin-1 Lys24-Pro477 Accession # A6NHN0 | |
| 25 μg | |
| Primary | |
| Detects human Otolin-1 in ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 839418 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C1q and tumor necrosis factor related protein 15, C1qTNF15, LOC131149, OTOL1, otolin 1, otolin-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 131149 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido