Learn More
Human Osteocalcin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
183.00€ - 538.00€
Especificaciones
Antígeno Osteocalcin
Clon 582014
Dilución Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
18776265
GREENER_CHOICE
Ver documentos
R&D Systems
MAB113272-SP
25 μg
183.00€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
18735865
GREENER_CHOICE
Ver documentos
R&D Systems
MAB113272-100
100 μg
538.00€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
Osteocalcin Monoclonal antibody specifically detects Osteocalcin in Human samples. It is validated for Flow Cytometry
Especificaciones
Especificaciones
Osteocalcin
Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
BGLAP, BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin
Osteocalcin/KLH containing peptide, Accession # P02818
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
582014
Flow Cytometry
Unconjugated
Mouse
Human
P02818
632
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.