missing translation for 'onlineSavingsMsg'
Learn More

Human NOV/CCN3 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18033402
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18033402 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18033402

Marca: R&D Systems AF1640

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Antibody has been used in 4 publications

NOV/CCN3 Polyclonal specifically detects NOV/CCN3 in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifica

Antígeno NOV/CCN3
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
N.º de referencia del gen P48745
Alias de gen CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog
Símbolos de los genes NOV
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human NOV/CCN3 Thr32-Met357 Accession # P48745
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4856
Especificidad de la prueba Detects human NOV/CCN3 in ELISAs and Western blots. In sandwich ELISAs, approximately 7% cross-reactivity with recombinant mouse NOV/CCN3 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.