missing translation for 'onlineSavingsMsg'
Learn More

Human Nectin-2/CD112 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18712099
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
Código de producto. Cantidad unitSize
18712099 25 μg 25 microgramos
18775439 100 μg 100 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18712099

Marca: R&D Systems MAB2229SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Nectin-2/CD112 Monoclonal specifically detects Nectin-2/CD112 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Nectin-2/CD112
Aplicaciones Flow Cytometry, Immunocytochemistry
Clasificación Monoclonal
Clon 610603
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen Q92692
Alias de gen CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HVEB, HVEBpoliovirus receptor-like 2, MPH, nectin 2, Nectin2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2, PRR2nectin-2, PVRL2, PVRR2poliovirus receptor related 2
Especie del huésped Mouse
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112 Gln32-Gly360 Accession # Q92692
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5819
Especificidad de la prueba Detects human Nectin-2/CD112 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.