missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TRANCE/TNFSF11/RANK L Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB6263-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse samples. It is validated for Western Blot.
Especificaciones
| TRANCE/TNFSF11/RANK L | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| AAC51762 | |
| TNFSF11 | |
| Mouse myeloma cell line NS0-derived recombinant human TRANCE/TNFSF11/RANK L Gly64-Asp245 Accession # AAC51762 | |
| 25 μg | |
| Primary | |
| Detects human TRANCE/TNFSF11/RANK L in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) TRANCE and approximately 25% cross-reactivity with recombinant human (rh) LIGHT is observed. No cross-reactivity with rh4-1BB Ligand, rhAPRIL, rhBAFF, rhEDA, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhOX40 LIgand, rhTNF-alpha , rhTRAIL, rhTWEAK, or rhVEGI is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 685809 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD254, CD254 antigen, ODF, OPGL, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANK L, RANKL, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8600 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido