missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse SOD1/Cu-Zn SOD Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18070822
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Código de producto. Cantidad unitSize
18070822 50 μg 50 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18070822

Marca: R&D Systems AF3418

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

SOD1/Cu-Zn SOD Polyclonal specifically detects SOD1/Cu-Zn SOD in Human, Mouse samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SOD1/Cu-Zn SOD
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.2 ug/mL, Simple Western 10 ug/mL
N.º de referencia del gen P00441
Alias de gen ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, CuZn SOD, Cu-Zn SOD, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, Ipo1, IPOA, SOD, SOD, cytosolic, SOD, Soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Símbolos de los genes SOD1
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human SOD1/Cu-Zn SOD Met1-Gln154 Accession # P00441
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6647
Especificidad de la prueba Detects human and mouse SOD1/Cu-Zn SOD in Western blots. Detects rat SOD1, but Catalog # AF3787 is recommended. In Western blots, less than 1% cross-reactivity with recombinant human SOD2 or SOD3 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human, Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.