Learn More
Human/Mouse PTEN Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Marca: R&D Systems MAB847
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PTEN Monoclonal specifically detects PTEN in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated.
Especificaciones
| PTEN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 μg/mL, Immunohistochemistry 8-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready, Knockout Validated | |
| P60484 | |
| PTEN | |
| E. coli-derived recombinant human PTEN Thr2-Val403 Accession # P60484 | |
| 100 μg | |
| Primary | |
| Detects human and mouse PTEN. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF, Immunoassay | |
| 217702 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5728 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |