missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse HAND2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF3876-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HAND2 Polyclonal specifically detects HAND2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| HAND2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BHLHA26, bHLHa26MGC125303, dHAND, DHAND2, Ehand2, FLJ16260, heart and neural crest derivatives expressed 2, heart, autonomic nervous system and neural crestderivatives-expressed protein 2, Hed, MGC125304, Th2, Thing2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9464 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P61296 | |
| HAND2 | |
| E. coli-derived recombinant human HAND2 Met1-Gln217 Accession # P61296 | |
| 25 μg | |
| Primary | |
| Detects human and mouse HAND2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human HAND1 is observed. | |
| Human, Mouse | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido