Learn More
Human/Mouse Doublecortin/DCX Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18759396
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18759396 25 μg 25 microgramos
18707516 100 μg 100 microgramos
2 options
Código de producto. 18759396
Marca: R&D Systems AF10025SP
Sheep Antibody
Doublecortin specifically detects Doublecortin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Doublecortin
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen O43602
Alias de gen DBCN, Doublecortex, doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, LISX, X-linked (doublecortin)
Especie del huésped Sheep
Inmunógeno E.coli-derived recombinant human Doublecortin/DCX, Met116-Thr293, Accession # O43602
Método de purificación Antigen Affinity-purified
Cantidad 25 μg
Primario o secundario Primary
ID de gen (Entrez) 1641
Especificidad de la prueba Detects human Doublecortin/DCX in direct ELISAs and Western blots. Detects mouse Doublecortin/DCX in immunohistochemistry.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human, Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Formulario Lyophilized
Isotype IgG