missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melusin Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF2740
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Melusin/ITGB1BP2 Polyclonal specifically detects Melusin/ITGB1BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
| Melusin/ITGB1BP2 | |
| Polyclonal | |
| Unconjugated | |
| Q9UKP3 | |
| ITGB1BP2 | |
| E. coli-derived recombinant human Melusin Ser2-Glu347 Accession # Q9UKP3 | |
| 100 μg | |
| Primary | |
| Detects human Melusin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| CHORDC3, integrin beta 1 binding protein (melusin) 2, integrin beta-1-binding protein 2, ITGB1BP, ITGB1BP2, Melusin, MGC119214 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 26548 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido