Learn More
Invitrogen™ Human LXR alpha (aa 318-329) Synthetic Peptide
Código de producto. 11514770
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Código de producto. 11514770
Marca: Invitrogen™ PEP114
Synthetic Peptide
Human LXR alpha (residues 318-329) Synthetic Peptide, PEP-114, from Invitrogen.
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiquitination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
TRUSTED_SUSTAINABILITY
Especificaciones
Gene ID (Entrez) 10062
Especie Human
Método de purificación purified
Número de acceso Q13133
Nombre común Human LXR alpha (aa 318-329)
Concentración 0.5 mg/mL
Contenido y almacenamiento -20°C. Avoid freeze-thaw cycles.
Descripción Synthetic Peptide
Formato Lyophilized
Formulación peptide with no preservative
Para utilizar con (aplicación) Control
Símbolo de gen NR1H3
Liofilizado Lyophilized
Conservante No preservative
Tipo de producto Peptide
Cantidad 50 μg
Estado normativo RUO - research use only
Secuencia C-D318-F-S-Y-N-R-E-D-F-A-K-A(329)