missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA1/CD85i/LIR-6 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems FAB30851R-100UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
LIR-6/LILRA1 Monoclonal specifically detects LIR-6/LILRA1 in Human samples. It is validated for Flow Cytometry.
Especificaciones
| LIR-6/LILRA1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O75019 | |
| LILRA1 | |
| Mouse myeloma cell line NS0-derived recombinant human LILRA1/CD85i/LIR-6 Pro17-Asn461 Accession # O75019 | |
| RUO | |
| 11024 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 586326 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD85 antigen-like family member I, CD85i, CD85i antigen, leukocyte immunoglobulin-like receptor subfamily A member 1, leukocyte immunoglobulin-like receptor subfamily A member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 1, LIR6, LIR-6, LIR-6Leukocyte immunoglobulin-like receptor 6, LIR6MGC126563 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human LILRA1/CD85i/LIR-6 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) ILT2 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido