Learn More
Invitrogen™ Human KIRREL3 ELISA Kit
Human KIRREL3 quantitates human KIRREL3 in serum, plasma, supernatant.
Marca:  Invitrogen™ EH288RBX10
Detalles adicionales : Peso : 2.50000kg
 Ver más versiones de este producto
Código de producto. 17844110
  • 4540.00€ / paquete de 10
Fecha estimada de envío: 28-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
Human KIRREL3 quantitates human KIRREL3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human KIRREL3.
The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
1500010O20Rik; 2900036G11Rik; KIAA1867; kin of IRRE like 3 (Drosophila); Kin of irregular chiasm-like protein 3; kin of IRRE-like protein 3; KIRRE; KIRREL3; membrane protein mKirre; mKIAA1867; mKirre; MRD4; NEPH2; nephrin-like 2; nephrin-like protein 2; PRO4502; Processed kin of IRRE-like protein 3; SST4; UNQ5923/PRO4502/PRO19814; x kin of IRRE like 3
0.21 ng/mL
ELISA
Colorimetric Microplate Reader
KIRREL3
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.21 to 50 ng/mL
Biotin
Supernatant
Q8IZU9
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL
KIRREL3
4 hr 45 min