Learn More
Human KIR2DL4/CD158d PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18768206
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
18768206 100 Tests 100 tests
18799328 25 Tests 25 tests
2 options
Código de producto. 18768206
Marca: R&D Systems FAB2238P100
Mouse Monoclonal Antibody has been used in 7 publications
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno KIR2DL4/CD158d
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 181703
Conjugado R-PE
Dilución Flow Cytometry 10 uL/10^6 cells
Formulación Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
N.º de referencia del gen Q99706
Alias de gen 103AS, 15.212, CD158 antigen-like family member D, CD158d, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor
Símbolos de los genes KIR2DL4
Especie del huésped Mouse
Inmunógeno BaF3 mouse pro-B cell line transfected with human KIR2DL4 Accession # Q99706
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3805
Especificidad de la prueba Stains human KIR2DL4 transfected BaF/3 cells. It does not recognize transfectants that express KIR2DL1, 2DL2, 2DL3, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Especies diana Human
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Formulario Purified
Isotype IgG2a