missing translation for 'onlineSavingsMsg'
Learn More

Human KIR2DL3/CD158b2 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. p-4715782
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
Código de producto. Cantidad unitSize
18765507 25 Tests 25 tests
18747626 100 Tests 100 tests
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18765507

Marca: R&D Systems FAB2014A025

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 3 publications

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Spezifikation

Antígeno KIR2DL3/CD158b2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 180701
Conjugado APC
Dilución Flow Cytometry 10 uL/10^6 cells
Formulación Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
N.º de referencia del gen P43628
Alias de gen CD158 antigen-like family member B2, CD158b2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, nkat2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Símbolos de los genes KIR2DL3
Especie del huésped Mouse
Inmunógeno BaF3 mouse pro-B cell line transfected with human KIR2DL3/CD158b2 Accession # P43628
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3804
Especificidad de la prueba Detects human KIR2DL3/CD158b2. Stains BaF3 cells transfected with human KIR2DL3 in flow cytometry. Does not stain BaF3 cells transfected with KIR2DL1, 2DL2, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Especies diana Human
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.