Learn More
Human Integrin beta 6 Biotinylated Antibody, R&D Systems™
Código de producto. 18149150
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
18149150 50 μg 50 microgramos
1 options
Código de producto. 18149150
Marca: R&D Systems BAF4155
Sheep Polyclonal Antibody
Integrin beta 6 Polyclonal specifically detects Integrin beta 6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Integrin beta 6
Aplicaciones Western Blot, Flow Cytometry, Immunocytochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Biotin
Dilución Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
N.º de referencia del gen P18564
Alias de gen integrin beta-6, integrin, beta 6, ITGB6
Símbolos de los genes ITGB6
Especie del huésped Sheep
Inmunógeno Mouse myeloma cell line NS0-derived recombinant human Integrin beta 6 Gly22-Asn707 Accession # P18564
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3694
Especificidad de la prueba Detects human Integrin beta 6 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse Integrin beta 6 is observed, approximately 10% cross-reactivity with recombinant human (rh) Integrin beta 3 is observed, and less than 5% cross-reactivity with rhIntegrin beta 5 is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG