Learn More
Human IL-6R alpha (Research Grade Tocilizumab Biosimilar) Antibody, R&D Systems™
Código de producto. 18754304
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18754304 100 μg 100 microgramos
18794404 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18754304 Proveedor R&D Systems N.º de proveedor MAB11482100
Human Monoclonal Antibody
IL-6R alpha Monoclonal antibody specifically detects IL-6R alpha in Human samples. It is validated for Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-6R alpha
Aplicaciones Flow Cytometry, ELISA, Blockade of Receptor-ligand Interaction
Clasificación Monoclonal
Clon Hu203
Conjugado Unconjugated
Dilución Flow Cytometry 0.25 μg/10^6 cells, ELISA, Blockade of Receptor-ligand Interaction 0.25 μg/10^6 cells
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
N.º de referencia del gen P08887
Alias de gen CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Especie del huésped Human
Inmunógeno E. coli-derived recombinant human IL-6R alpha, Arg387-Arg468, Accession # P08887
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3570
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.