Learn More
Human IL-6R alpha Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18752679
Change view
Click to view available options
Cantidad:
1 mg
100 μg
25 μg
500 ug
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
500 microgramos
18752679 25 μg 25 microgramos
18730387 100 μg 100 microgramos
18120351 500 ug 500 microgramos
18179060 1 mg 1 miligramo
4 options
Código de producto. 18752679
Marca: R&D Systems MAB2271RSP
Mouse Monoclonal Antibody
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-6R alpha
Aplicaciones Immunocytochemistry
Clasificación Monoclonal
Clon 556704R
Conjugado Unconjugated
Dilución Immunocytochemistry 3-25 ug/mL
Formulación Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P08887
Alias de gen CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Símbolos de los genes IL6R
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human IL-6 R alpha Arg387-Arg468 Accession # P08887
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3570
Especificidad de la prueba Detects human IL-6 R alpha in direct ELISAs.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only