Learn More
Human IL-33 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18740599
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
18740599 100 μg 100 microgramos
18715419 25 μg 25 microgramos
2 options
Código de producto. 18740599
Marca: R&D Systems MAB36254100
Goat Monoclonal Antibody
IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-33
Aplicaciones Western Blot, Immunohistochemistry, Neutralization
Clasificación Monoclonal
Clon 40015D
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Neutralization 0.75-3 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen O95760
Alias de gen C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Símbolos de los genes IL33
Especie del huésped Goat
Inmunógeno E. coli-derived recombinant human IL-33 Ser112-Thr270 Accession # O95760
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 90865
Especificidad de la prueba Detects human IL-33 in direct ELISAs.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG