Learn More
Human IL-22 R alpha 1 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18041742
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
18041742 100 μg 100 microgramos
1 options
Código de producto. 18041742
Marca: R&D Systems AF2770
Goat Polyclonal Antibody has been used in 4 publications
IL-22R alpha 1 Polyclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IL-22R alpha 1
Aplicaciones Neutralization
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Neutralization 0.2-1 ug/mL
N.º de referencia del gen Q8N6P7
Alias de gen CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Símbolos de los genes IL22RA1
Especie del huésped Goat
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7
Método de purificación Affinity Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 58985
Especificidad de la prueba Detects human IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human IL-20 R beta is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG