Learn More
STEMCELL Technologies Kit de ELISA para IL-2 humano
Código de producto. 17148011
Encompass_Preferred
Change view
Click to view available options
Cantidad:
2 placas
Tamaño de la unidad:
paquete de 2
17148011 2 placas paquete de 2
1 options
Código de producto. 17148011
Marca: STEMCELL Technologies 02006
For detection and measurement of human interleukin 2
  • The Human Interleukin 2 (IL-2) ELISA Kit is designed for the quantitative detection and measurement of human IL-2 in biological fluids such as serum, plasma, and cell culture supernatants
  • IL-2 is primarily secreted by activated CD4+ and CD8+ T cells
  • IL-2 is a major regulator of T cell development, contributing to T cell growth and expansion, and the development of regulatory T cells
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Descripción B Cells; Dendritic Cells; T Cells, CD4+; T Cells, CD8+
Para utilizar con (aplicación) ELISA
Cantidad 2 placas