Learn More
STEMCELL Technologies Kit de ELISA para IL-2 humano
Descripción
- The Human Interleukin 2 (IL-2) ELISA Kit is designed for the quantitative detection and measurement of human IL-2 in biological fluids such as serum, plasma, and cell culture supernatants
- IL-2 is primarily secreted by activated CD4+ and CD8+ T cells
- IL-2 is a major regulator of T cell development, contributing to T cell growth and expansion, and the development of regulatory T cells
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Especificaciones
| Descripción | B Cells; Dendritic Cells; T Cells, CD4+; T Cells, CD8+ |
| Para utilizar con (aplicación) | ELISA |
| Cantidad | 2 placas |