missing translation for 'onlineSavingsMsg'
Learn More

Human IL-12/IL-23 p40 MAb (Clone 169516), R&D Systems™
GREENER_CHOICE

Código de producto. p-7067944
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
Código de producto. Cantidad unitSize
18716038 25 μg 25 microgramos
18759128 100 μg 100 microgramos
18747139 500 μg 500 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18716038

Marca: R&D Systems MAB6091SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 2 publications

IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-12/IL-23 p40
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 169516
Conjugado Unconjugated
Dilución Western Blot 1 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P29460
Alias de gen CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40
Símbolos de los genes IL12B
Especie del huésped Mouse
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-23 p19/p40 heterodimer Ile23-Ser328 Accession # P29460
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3593
Especificidad de la prueba Detects human IL-12/IL-23 p40 subunit in direct ELISAs and Western blots. Recognizes IL-12 and IL-23 heterodimers as well as p40 homodimers. Shows 100% cross-reactivity with porcine IL-12/IL-23 p40 and no cross-reactivity with mouse IL-12/IL-23 p40.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.