missing translation for 'onlineSavingsMsg'
Learn More

Human IGSF8/CD316 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18775838
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18775838 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18775838 Proveedor R&D Systems N.º de proveedor FAB3117U100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGSF8/CD316 Monoclonal specifically detects IGSF8/CD316 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IGSF8/CD316
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 2587A
Conjugado Alexa Fluor 350
Dilución Flow Cytometry 0.25-1 ug/10^6 cells
Formulación Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
N.º de referencia del gen Q969P0
Alias de gen CD316, CD316 antigen, CD81P3, CD81P3LIR-D1, EWI2, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRL, PGRLimmunoglobulin superfamily member 8
Especie del huésped Rabbit
Inmunógeno Human embryonic kidney cell, HEK293-derived human IGSF8/CD316 protein, Arg28-Thr579, Accession # Q969P0
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 93185
Especies diana Human
Contenido y almacenamiento Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.