missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG4 Polyclonal Antibody

Descripción
Human IgG4 Polyclonal Antibody for Western Blot, IHC (P)
Human IgG4, also known as human immunoglobulin G4, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG4 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG4 is also involved in other immune responses, such as immune tolerance and suppression of inflammation. Human IgG4 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Especificaciones
Especificaciones
| Antígeno | Human IgG4 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | IGHG4 |
| N.º de referencia del gen | P01861 |
| Alias de gen | IGHG4; Immunoglobulin G; Immunoglobulin G4; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG |
| Símbolos de los genes | IGHG4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?