Learn More
STEMCELL Technologies Human IgG ELISA Antibody Pair Kit
Código de producto. 17157991
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 kit para 6 placas
Tamaño de la unidad:
1 set
17157991 1 kit para 6 placas 1 set
1 options
Código de producto. 17157991
Marca: STEMCELL Technologies 01994
For detection and measurement of human immunoglobulin G
  • The Human Immunoglobulin G (IgG) ELISA Antibody Pair Kit is intended for those who want the flexibility of setting up their own ELISA assay
  • This kit includes capture and detection antibodies and an IgG standard
  • It is designed for the quantitative detection and measurement of total human IgG in biological fluids such as serum, plasma, and cell culture supernatants
  • IgG is the major immunoglobulin found in blood, lymph fluid, cerebrospinal fluid, and peritoneal fluid
  • IgG plays a key role in the humoral immune response, plays a critical step in the opsonization process by interacting with the Fcγ receptor on macrophages, neutrophils, and natural killer cells, and is able to activate the complement system
  • NOTE: This kit includes sufficient reagents for 6 x 96-well ELISA plates (Catalog #38019)
  • ELISA plates and pNPP Substrate (Catalog #01917) are required for use with the Human IgG ELISA Antibody Pair Kit and are available for purchase separately
Especificaciones
Descripción B Cells; Hybridomas
Cantidad 1 kit para 6 placas