Learn More
STEMCELL Technologies Human IgE ELISA Kit
Código de producto. p-7217812
Encompass_Preferred
Click to view available options
Cantidad:
2 placas
Tamaño de la unidad:
paquete de 2
Código de producto. 17158041
Marca: STEMCELL Technologies 02032
For detection and measurement of human immunoglobulin E
  • The Human Immunoglobulin E (IgE) ELISA Kit is designed for the quantitative detection and measurement of human IgE in biological fluids such as serum, plasma, and cell culture supernatants
  • IgE is found in the lungs, skin, and mucous membranes
  • IgE plays a role in allergic reactions by stimulating degranulation of eosinophils, mast cells, and basophils through interaction with the Fc epsilon Receptor I (FcεRI)
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the immunoglobulin
  • The captured immunoglobulin is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of immunoglobulin in the sample
Especificaciones
Descripción B Cells; Hybridomas
Cantidad 2 placas
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.