Learn More
STEMCELL Technologies Human IgE ELISA Kit
Click to view available options
Cantidad:
2 placas
Tamaño de la unidad:
paquete de 2
Descripción
- The Human Immunoglobulin E (IgE) ELISA Kit is designed for the quantitative detection and measurement of human IgE in biological fluids such as serum, plasma, and cell culture supernatants
- IgE is found in the lungs, skin, and mucous membranes
- IgE plays a role in allergic reactions by stimulating degranulation of eosinophils, mast cells, and basophils through interaction with the Fc epsilon Receptor I (FcεRI)
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the immunoglobulin
- The captured immunoglobulin is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of immunoglobulin in the sample
Especificaciones
Especificaciones
| Descripción | B Cells; Hybridomas |
| Cantidad | 2 placas |