missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgA Polyclonal Antibody

Descripción
Reconstitute at 0.2 mg/mL in sterile PBS.
Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
Especificaciones
Especificaciones
| Antígeno | Human IgA |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with trehalose and no preservative |
| génica | IGHA1 |
| N.º de referencia del gen | P01876, P01877 |
| Alias de gen | A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker) |
| Símbolos de los genes | IGHA1, IGHA2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?