Learn More
Human IFN-gamma R1/CD119 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18778438
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18778438 500 μg 500 microgramos
18762848 25 μg 25 microgramos
18715158 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18778438 Proveedor R&D Systems N.º de proveedor MAB6732500
Mouse Monoclonal Antibody has been used in 4 publications
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IFN-gamma R1/CD119
Aplicaciones Neutralization
Clasificación Monoclonal
Clon GIR208
Conjugado Unconjugated
Dilución Neutralization 0.03-0.15 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias de gen AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Símbolos de los genes IFNGR1
Especie del huésped Mouse
Inmunógeno Human placenta-derived IFN-gamma R1/CD119
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3459
Especificidad de la prueba Detects human IFN-gamma R1/CD119 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta is observed.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Supernatant
Isotype IgG1