missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hemoglobin zeta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB7708-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Hemoglobin zeta Monoclonal specifically detects Hemoglobin zeta in Human samples. It is validated for Western Blot, Immunocytochemistry.
Especificaciones
| Hemoglobin zeta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P02008 | |
| HBZ | |
| E. coli-derived recombinant human Hemoglobin zeta Met1-Arg142 Accession # P02008 | |
| 25 μg | |
| Primary | |
| Detects human Hemoglobin zeta in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 778633 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HBAZ, HBZ, HBZ2, hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, zeta, zeta-globin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3050 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido