missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HDGF Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF1606
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HDGF Polyclonal specifically detects HDGF in Human samples. It is validated for Western Blot.
Especificaciones
| HDGF | |
| Polyclonal | |
| Unconjugated | |
| P51858 | |
| HDGF | |
| E. coli-derived recombinant human HDGF Ser2-Lys240 (Pro101Lys) Accession # P51858 | |
| 100 μg | |
| Primary | |
| Detects human HDGF in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| FLJ96580, hepatoma-derived growth factor, hepatoma-derived growth factor (high-mobility group protein 1-like), High mobility group protein 1-like 2, HMG-1L2, HMG1L2DKFZp686J1764, HuHGF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3068 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido