missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glut1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB14181
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for Western Blot, Simple Western.Información de órdenes
The product is shipped at ambient temperature.
Especificaciones
| Glut1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, SLC2A1, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6513 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
| Western Blot | |
| 202921 | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL | |
| AAA52571 | |
| SLC2A1 | |
| NS0 mouse myeloma cell line transfected with human Glut1 Met1-Val492 Accession # AAA52571 | |
| 100 μg | |
| Primary | |
| Detects human Glut1 in ELISAs. | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido