missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-3 Antibody, R&D Systems™

Beskrivelse
Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Tekniske data
Tekniske data
| Antígeno | Galectin-3 |
| Aplicaciones | Western Blot, Immunohistochemistry, ELISA |
| Clasificación | Monoclonal |
| Clon | 194805 |
| Concentración | LYOPH |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| N.º de referencia del gen | P17931.5 |
| Alias de gen | AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?