missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FAM82A1 Synthetic Peptide
Descripción
FAM82A1 Synthetic Peptide, PEP-1052, from Invitrogen.
The human protein FAM82A1, also known as regulator of microtubule dynamics 2 (RMD2), was one of three proteins identified as related to a family of microtubule-associated proteins in C. elegans. FAM82A1 contains multiple coiled-coil domains and localize to the spindle microtubules and spindle poles during cell division. During interphase, RMD2 localizes to the cytoplasm with protein observed in the microtubule lattice and perinuclear region.
Specifications
Specifications
| Gene ID (Entrez) | 151393 |
| Especie | Human |
| Método de purificación | purified |
| Número de acceso | Q96LZ7 |
| Nombre común | Human FAM82A1 |
| Concentración | 200 μg/mL |
| Contenido y almacenamiento | -20°C |
| Descripción | Synthetic Peptide |
| Formato | Liquid |
| Formulación | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?