Learn More
Invitrogen™ Human Estrogen Receptor alpha (aa 21-32) Synthetic Peptide
Código de producto. 11594690
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
11594690 50 μg 50 microgramos
1 options
Código de producto. 11594690 Proveedor Invitrogen™ N.º de proveedor PEP037
Synthetic Peptide
Human Estrogen Receptor A (ERa) (Residues 21-32) Synthetic Peptide, PEP-037, from Invitrogen.
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.
TRUSTED_SUSTAINABILITY
Especificaciones
Gene ID (Entrez) 2099
Especie Human
Método de purificación purified
Número de acceso P03372
Nombre común Human Estrogen Receptor alpha (aa 21-32)
Concentración 0.5 mg/mL
Contenido y almacenamiento -20°C. Avoid freeze-thaw cycles.
Descripción Synthetic Peptide
Formato Lyophilized
Formulación peptide with no preservative
Para utilizar con (aplicación) Control
Símbolo de gen ESR1
Liofilizado Lyophilized
Conservante No preservative
Tipo de producto Peptide
Cantidad 50 μg
Estado normativo RUO - research use only
Secuencia N21 - E - L - E - P - L - N - R - P - Q - L - K32 - (C)