Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18749427
Change view
Click to view available options
Cantidad:
100 μg
25 μg
500 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
18749427 25 μg 25 microgramos
18775558 100 μg 100 microgramos
18782579 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 18749427 Proveedor R&D Systems N.º de proveedor MAB287SP
Mouse Monoclonal Antibody has been used in 5 publications
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Erythropoietin/EPO
Aplicaciones Neutralization
Clasificación Monoclonal
Clon 9C21D11
Conjugado Unconjugated
Dilución Neutralization 0.5-1.5 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P01588
Alias de gen ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Símbolos de los genes EPO
Especie del huésped Mouse
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human Erythropoietin/EPO, Accession # P01588
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2056
Especificidad de la prueba Detects human Erythropoietin/EPO.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG1