missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Endoglin/CD105 Antibody, R&D Systems™

Descripción
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Especificaciones
Especificaciones
| Antígeno | Endoglin/CD105 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 166713 |
| Concentración | LYOPH |
| Conjugado | Unconjugated |
| Dilución | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| N.º de referencia del gen | P17813 |
| Alias de gen | CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 |
| Mostrar más |
For Research Use Only
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?