missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DR3/TNFRSF25 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF943
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blot.
Especificaciones
| DR3/TNFRSF25 | |
| Polyclonal | |
| Unconjugated | |
| Q93038 | |
| TNFRSF25 | |
| Mouse myeloma cell line NS0-derived recombinant human DR3/TNFRSF25 Gln25-Phe201 Accession # Q93038 | |
| 100 μg | |
| Primary | |
| Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse DR3 is observed, and less than 1% cross-reactivity with recombinant human (rh) Fas, rhTNF RI, rhDR6, rh4-1BB, rhCD27, rhCD30, rhCD40, rhGITR, rhHVEM, rhTNF RII and rhOPG is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8718 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido