missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Desmin ELISA Kit

Human Desmin quantitates human Desmin in serum, plasma, supernatant.

Marca:  Invitrogen™ EH152RBX10

Detalles adicionales : Peso : 2.50000kg

 Ver más versiones de este producto

Código de producto. 17814080

  • 4540.00€ / paquete de 10

Fecha estimada de envío: 27-04-2023
para ver el stock
Añadir a la cesta

Descripción

Descripción

Human Desmin quantitates human Desmin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Desmin.

Desmin (approximately 53 kDa) exhibits a high degree of tissue specificity, its expression being predominantly confined to all types of muscle cells (cardiac, skeletal and smooth muscle). Regulation of desmin expression is stage and tissue-specific, since it is induced during terminal development of, for example, skeletal muscle cell differentiation. In skeletal en cardiac muscle cells desmin is localized in the Z-disk region and at the intercalated disk. The expression pattern of desmin in smooth muscle is much more heterogenous. Coexpression of vimentin and desmin has been observed in tumors derived from muscle tissue, i.e. rhabdomyosarcomas and leiomyosarcomas. Furthermore, during myocard dysfunction dramatic changes in the distribution of desmin have been observed.
Especificaciones

Especificaciones

CMD1I; CSM1; CSM2; Des; desmin; desmin-like protein; FLJ12025; FLJ39719; FLJ41013; FLJ41793; I79_018932; intermediate filament protein; LGMD2R; MFM1; muscle-specific intermediate filament desmin; muscle-specific intermediate filament protein; mutant desmin p.K241E; SCPNK; similar to desmin
2 ng/mL
ELISA
Colorimetric Microplate Reader
DES
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
2 to 500 ng/mL
Biotin
Supernatant
P17661
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Desmin
4 hr 45 min
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales