missing translation for 'onlineSavingsMsg'
Learn More

Human DEK Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18152911
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
18152911 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18152911

Marca: R&D Systems MAB70201

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DEK Monoclonal specifically detects DEK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno DEK
Aplicaciones Immunohistochemistry, Immunocytochemistry
Clasificación Monoclonal
Clon 715511
Concentración LYOPH
Conjugado Unconjugated
Dilución Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P35659
Alias de gen D6S231EDEK oncogene (DNA binding), DEK oncogene, protein DEK
Símbolos de los genes DEK
Especie del huésped Mouse
Inmunógeno Human human embryonic kidney cell line HEK293EBNA-derived recombinant human DEK Val282-Ser375 Accession # P35659
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7913
Especificidad de la prueba Detects human DEK in direct ELISAs.
Reconstitución Sterile PBS to a final concentration of 0.5 mg/mL.
Especies diana Human
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.