Learn More
Invitrogen™ Human Cystatin A ELISA Kit
Human Cystatin A quantitates human Cystatin A in serum, plasma, supernatant.
Marca: Invitrogen™ EH140RBX10
3041.80 EUR hasta el 2023-03-27
Use el código promocional "20585" para aplicar la oferta.
Detalles adicionales : Peso : 2.50000kg
Descripción
Human Cystatin A quantitates human Cystatin A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cystatin A.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.Especificaciones
AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; Stefin-A; STF1; STFA | |
6.5 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CSTA | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
6.5 to 1,500 pg/mL | |
Biotin | |
Supernatant | |
P01040 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
Cystatin A | |
4 hr 45 min |